Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83E
Entrez Gene ID
54854
Gene Name
family with sequence similarity 83 member E
Gene Aliases
-
Chromosome Location
Chr.19: 48600600 - 48614841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017708.3 NP_060178.2 1 - 2 534 64
XM_005259019.1 XP_005259076.1 2 - 3 1858 64
XM_011527049.2 XP_011525351.1 2 - 3 1843 64
GenBank mRNA BC111970.1 - 1 - 2 534 64
BC111972.1 - 1 - 2 534 64

View Full Product Details