The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000788.2 | NP_000779.1 | 2 - 3 | 495 | 111 |
GenBank mRNA | AK303292.1 | - | 2 - 3 | 571 | 111 |
AK313523.1 | - | 2 - 3 | 446 | 111 | |
BC103764.1 | - | 2 - 3 | 406 | 111 | |
BC114617.1 | - | 2 - 3 | 366 | 111 | |
BI601939.1 | - | 2 - 3 | 496 | 111 | |
BT019941.1 | - | 2 - 3 | 207 | 111 | |
BT019942.1 | - | 2 - 3 | 207 | 111 | |
CD014016.1 | - | 2 - 3 | 496 | 111 | |
CR536527.1 | - | 2 - 3 | 207 | 111 | |
CR541876.1 | - | 2 - 3 | 207 | 111 | |
JF806429.1 | - | 2 - 3 | 247 | 111 | |
M60527.1 | - | 2 - 3 | 366 | 111 |