The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014244.4 | NP_055059.2 | 18 - 19 | 2859 | 83 |
XM_017010063.1 | XP_016865552.1 | 17 - 18 | 2295 | 83 | |
GenBank mRNA | AJ003125.1 | - | 18 - 19 | 2757 | 83 |
CO247372.1 | - | 4 - 5 | 492 | 83 |