The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014244.4 | NP_055059.2 | 19 - 20 | 3061 | 64 |
XM_017010063.1 | XP_016865552.1 | 18 - 19 | 2497 | 64 | |
GenBank mRNA | AJ003125.1 | - | 19 - 20 | 2959 | 64 |