The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006334.3 | NP_006325.1 | 3 - 4 | 851 | 85 |
GenBank mRNA | AI656802.1 | - | 1 - 2 | 85 | |
D82343.1 | - | 3 - 4 | 686 | 85 |