Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPTM1
Entrez Gene ID
1209
Gene Name
CLPTM1, transmembrane protein
Gene Aliases
-
Chromosome Location
Chr.19: 44954585 - 44993346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282175.1 NP_001269104.1 9 - 10 1155 100
NM_001282176.1 NP_001269105.1 9 - 10 1179 100
NM_001294.3 NP_001285.1 9 - 10 1306 100
GenBank mRNA AF037339.1 - 9 - 10 1149 100
AK074935.1 - 9 - 10 1150 100
AK075262.1 - 9 - 10 1149 100
AK223553.1 - 9 - 10 1150 100
AK293314.1 - 9 - 10 1171 100
AK304480.1 - 9 - 10 1138 100
AK316094.1 - 9 - 10 1175 100
BC004865.2 - 9 - 10 1185 100
BC012359.1 - 9 - 10 1139 100
BT007262.1 - 9 - 10 1134 100
KU177986.1 - 9 - 10 1134 100
KU177987.1 - 9 - 10 1134 100

View Full Product Details