Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICAL3
Entrez Gene ID
57553
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases
MICAL-3
Chromosome Location
Chr.22: 17787649 - 18024559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015241.2 NP_056056.2 28 - 29 5910 124
XM_005261262.3 XP_005261319.1 28 - 29 5740 124
XM_011546137.2 XP_011544439.1 35 - 36 6310 124
XM_011546139.2 XP_011544441.1 35 - 36 6310 124
XM_011546142.2 XP_011544444.1 34 - 35 6259 124
XM_017028865.1 XP_016884354.1 35 - 36 6828 124
XM_017028866.1 XP_016884355.1 35 - 36 6634 124
XM_017028867.1 XP_016884356.1 35 - 36 7751 124
XM_017028868.1 XP_016884357.1 35 - 36 6480 124
XM_017028869.1 XP_016884358.1 35 - 36 6225 124
XM_017028870.1 XP_016884359.1 35 - 36 6226 124
XM_017028871.1 XP_016884360.1 35 - 36 6307 124
XM_017028872.1 XP_016884361.1 34 - 35 6259 124
XM_017028873.1 XP_016884362.1 34 - 35 6247 124
XM_017028874.1 XP_016884363.1 34 - 35 6226 124
XM_017028875.1 XP_016884364.1 34 - 35 6202 124
XM_017028876.1 XP_016884365.1 34 - 35 6172 124
XM_017028877.1 XP_016884366.1 33 - 34 6109 124
XM_017028878.1 XP_016884367.1 32 - 33 5995 124
XM_017028879.1 XP_016884368.1 30 - 31 5854 124
XM_017028880.1 XP_016884369.1 30 - 31 6177 124
XM_017028881.1 XP_016884370.1 30 - 31 6024 124
XM_017028882.1 XP_016884371.1 29 - 30 5791 124
XM_017028883.1 XP_016884372.1 34 - 35 5973 124
XM_017028884.1 XP_016884373.1 31 - 32 6303 124
XM_017028885.1 XP_016884374.1 15 - 16 3895 124
XM_017028886.1 XP_016884375.1 15 - 16 3743 124
GenBank mRNA AB020626.1 - 8 - 9 2518 124
AK126925.1 - 1 - 2 796 124
BC006562.2 - 3 - 4 684 124

View Full Product Details