The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015241.2 | NP_056056.2 | 29 - 30 | 6004 | 80 |
XM_005261262.3 | XP_005261319.1 | 29 - 30 | 5834 | 80 | |
XM_011546139.2 | XP_011544441.1 | 36 - 37 | 6404 | 80 | |
XM_011546142.2 | XP_011544444.1 | 35 - 36 | 6353 | 80 | |
GenBank mRNA | AB020626.1 | - | 9 - 10 | 2612 | 80 |
AK126925.1 | - | 2 - 3 | 890 | 80 | |
BC006562.2 | - | 4 - 5 | 778 | 80 |