Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 27 - 28 3364 100
NM_001254719.1 NP_001241648.1 26 - 27 3310 100
NM_001254720.1 NP_001241649.1 25 - 26 3274 100
NM_001254721.1 NP_001241650.1 25 - 26 3253 100
NM_001254722.1 NP_001241651.1 24 - 25 3232 100
NM_001254723.1 NP_001241652.1 25 - 26 3271 100
NM_002465.3 NP_002456.2 28 - 29 3385 100
NM_206819.2 NP_996555.1 28 - 29 3385 100
NM_206820.2 NP_996556.1 27 - 28 3364 100
NM_206821.2 NP_996557.1 26 - 27 3310 100
XM_005268876.4 XP_005268933.1 27 - 28 3319 100
XM_006719405.3 XP_006719468.1 29 - 30 3300 100
XM_006719406.3 XP_006719469.1 29 - 30 3300 100
XM_006719407.3 XP_006719470.1 28 - 29 3373 100
XM_006719408.3 XP_006719471.1 28 - 29 3246 100
XM_006719409.3 XP_006719472.1 27 - 28 3326 100
XM_006719410.3 XP_006719473.1 29 - 30 3300 100
XM_006719411.3 XP_006719474.1 27 - 28 3326 100
XM_017019315.1 XP_016874804.1 28 - 29 3243 100
XM_017019316.1 XP_016874805.1 29 - 30 3300 100
XM_017019317.1 XP_016874806.1 26 - 27 3272 100
XM_017019318.1 XP_016874807.1 27 - 28 3189 100
XM_017019319.1 XP_016874808.1 26 - 27 3269 100
XM_017019320.1 XP_016874809.1 25 - 26 3368 100
XM_017019321.1 XP_016874810.1 25 - 26 3215 100
XM_017019322.1 XP_016874811.1 23 - 24 3257 100
GenBank mRNA AK057284.1 - 8 - 9 1429 100
AK295727.1 - 24 - 25 3192 100
AK296681.1 - 27 - 28 3328 100
AK303401.1 - 25 - 26 3231 100
AL831993.2 - 28 - 29 3320 100
AL832000.2 - 28 - 29 3320 100
AL832332.2 - 28 - 29 3320 100
AL832350.2 - 28 - 29 3320 100
AL832354.2 - 28 - 29 3323 100
AL832456.2 - 28 - 29 3320 100
AL834249.2 - 28 - 29 3320 100
BC092418.1 - 26 - 27 3257 100
BC117217.1 - 24 - 25 3167 100
BC143495.1 - 25 - 26 3188 100
BC143502.1 - 24 - 25 3167 100
BC143503.1 - 26 - 27 3245 100
BC143504.1 - 25 - 26 3209 100
BX649073.2 - 29 - 30 3415 100
X66276.1 - 27 - 28 3311 100
X73114.1 - 26 - 27 3251 100

View Full Product Details