The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177948.1 | NP_001171419.1 | 3 - 4 | 323 | 70 |
NM_001177949.1 | NP_001171420.1 | 3 - 4 | 338 | 70 | |
NM_153694.4 | NP_710161.1 | 3 - 4 | 334 | 70 | |
XM_005268922.4 | XP_005268979.3 | 3 - 4 | 584 | 70 | |
XM_005268924.1 | XP_005268981.1 | 3 - 4 | 322 | 70 | |
XM_005268925.1 | XP_005268982.1 | 3 - 4 | 327 | 70 | |
XM_005268926.3 | XP_005268983.1 | 3 - 4 | 306 | 70 | |
XM_005268927.2 | XP_005268984.1 | 3 - 4 | 295 | 70 | |
XM_011538421.2 | XP_011536723.2 | 3 - 4 | 506 | 70 | |
XM_017019368.1 | XP_016874857.1 | 3 - 4 | 581 | 70 | |
GenBank mRNA | AF492003.1 | - | 3 - 4 | 279 | 70 |
AF517774.1 | - | 3 - 4 | 277 | 70 | |
BC062662.1 | - | 3 - 4 | 329 | 70 | |
BQ428625.1 | - | 3 - 4 | 339 | 70 |