Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCP3
Entrez Gene ID
50511
Gene Name
synaptonemal complex protein 3
Gene Aliases
COR1, RPRGL4, SCP3, SPGF4
Chromosome Location
Chr.12: 101728648 - 101739722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177948.1 NP_001171419.1 4 - 5 355 97
NM_001177949.1 NP_001171420.1 4 - 5 370 97
NM_153694.4 NP_710161.1 4 - 5 366 97
XM_005268922.4 XP_005268979.3 4 - 5 616 97
XM_005268924.1 XP_005268981.1 4 - 5 354 97
XM_005268925.1 XP_005268982.1 4 - 5 359 97
XM_005268926.3 XP_005268983.1 4 - 5 338 97
XM_005268927.2 XP_005268984.1 4 - 5 327 97
XM_011538421.2 XP_011536723.2 4 - 5 538 97
XM_017019368.1 XP_016874857.1 4 - 5 613 97
GenBank mRNA AF492003.1 - 4 - 5 311 97
AF517774.1 - 4 - 5 309 97
AK125930.1 - 2 - 3 1021 257
BC062662.1 - 4 - 5 361 97
BQ428625.1 - 4 - 5 371 97

View Full Product Details