The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191002.1 | NP_001177931.1 | 1 - 2 | 250 | 98 |
NM_004832.2 | NP_004823.1 | 1 - 2 | 250 | 98 | |
GenBank mRNA | AF212303.1 | - | 1 - 2 | 57 | 98 |
AK313680.1 | - | 1 - 2 | 68 | 98 | |
BC000127.1 | - | 1 - 2 | 60 | 98 | |
BX537431.1 | - | 1 - 2 | 178 | 98 | |
CN283947.1 | - | 1 - 2 | 53 | 98 | |
DT220068.1 | - | 1 - 2 | 285 | 98 | |
EU794619.1 | - | 1 - 2 | 57 | 98 |