The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304743.1 | NP_001291672.1 | 11 - 12 | 3165 | 105 |
NM_014720.3 | NP_055535.2 | 11 - 12 | 3165 | 105 | |
XM_011540401.2 | XP_011538703.1 | 10 - 11 | 1821 | 105 | |
GenBank mRNA | AB002804.1 | - | 11 - 12 | 2836 | 105 |
AF273048.1 | - | 11 - 12 | 2638 | 105 | |
AK302855.1 | - | 11 - 12 | 2598 | 105 | |
BC111565.1 | - | 11 - 12 | 2635 | 105 | |
D86959.1 | - | 11 - 12 | 3115 | 105 |