The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258206.1 | NP_001245135.1 | 11 - 12 | 1230 | 72 |
NM_001258207.1 | NP_001245136.1 | 10 - 11 | 1134 | 72 | |
NM_005428.3 | NP_005419.2 | 11 - 12 | 1230 | 72 | |
XM_005259642.1 | XP_005259699.1 | 11 - 12 | 1190 | 72 | |
GenBank mRNA | AK225710.1 | - | 11 - 12 | 1184 | 72 |
AK313307.1 | - | 11 - 12 | 1172 | 72 | |
BC013361.2 | - | 11 - 12 | 1010 | 72 | |
X83931.1 | - | 3 - 4 | 196 | 72 |