The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144578.3 | NP_653179.1 | 2 - 3 | 185 | 84 |
XM_017021789.1 | XP_016877278.1 | 2 - 3 | 221 | 84 | |
GenBank mRNA | AK025580.1 | - | 2 - 3 | 148 | 84 |
AK315501.1 | - | 2 - 3 | 182 | 84 | |
AL834285.1 | - | 2 - 3 | 168 | 84 | |
BC015621.2 | - | 2 - 3 | 146 | 84 |