The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001312660.1 | NP_001299589.1 | 5 - 6 | 685 | 102 |
NM_145870.2 | NP_665877.1 | 4 - 5 | 499 | 102 | |
GenBank mRNA | AJ001838.1 | - | 4 - 5 | 320 | 102 |
AK315154.1 | - | 5 - 6 | 576 | 102 | |
AK315609.1 | - | 4 - 5 | 343 | 102 | |
BC001453.1 | - | 4 - 5 | 375 | 102 | |
BI559916.1 | - | 5 - 6 | 594 | 102 | |
BI829992.1 | - | 5 - 6 | 565 | 102 | |
CR456987.1 | - | 3 - 4 | 217 | 102 | |
U86529.1 | - | 4 - 5 | 320 | 102 |