The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017572.3 | NP_060042.2 | 8 - 9 | 839 | 70 |
NM_199054.2 | NP_951009.1 | 8 - 9 | 839 | 70 | |
GenBank mRNA | AF125532.1 | - | 7 - 8 | 616 | 70 |
AF237775.1 | - | 7 - 8 | 630 | 70 | |
AF237776.1 | - | 7 - 8 | 630 | 70 | |
AK293742.1 | - | 6 - 7 | 498 | 70 | |
AK303393.1 | - | 5 - 6 | 509 | 70 | |
AL137565.1 | - | 4 - 5 | 301 | 70 | |
BC073140.1 | - | 8 - 9 | 819 | 70 |