The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018218.2 | NP_060688.1 | 11 - 12 | 1580 | 140 |
XM_006712612.3 | XP_006712675.1 | 11 - 12 | 1598 | 140 | |
XM_011511396.2 | XP_011509698.1 | 11 - 12 | 1598 | 140 | |
XM_011511397.2 | XP_011509699.1 | 12 - 13 | 1672 | 140 | |
XM_011511398.2 | XP_011509700.1 | 11 - 12 | 1598 | 140 | |
XM_011511399.2 | XP_011509701.1 | 10 - 11 | 1399 | 140 | |
XM_011511400.2 | XP_011509702.1 | 10 - 11 | 1383 | 140 | |
XM_011511401.2 | XP_011509703.1 | 11 - 12 | 1598 | 140 | |
XM_017004426.1 | XP_016859915.1 | 12 - 13 | 1672 | 140 | |
XM_017004427.1 | XP_016859916.1 | 5 - 6 | 600 | 140 | |
GenBank mRNA | AJ583821.2 | - | 11 - 12 | 1580 | 140 |
AK001647.1 | - | 11 - 12 | 1542 | 140 | |
AK297821.1 | - | 5 - 6 | 668 | 140 |