Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP40
Entrez Gene ID
55230
Gene Name
ubiquitin specific peptidase 40
Gene Aliases
-
Chromosome Location
Chr.2: 233475519 - 233566789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018218.2 NP_060688.1 28 - 29 3444 89
XM_006712612.3 XP_006712675.1 28 - 29 3462 89
XM_011511396.2 XP_011509698.1 28 - 29 3474 89
XM_011511397.2 XP_011509699.1 29 - 30 3548 89
XM_011511399.2 XP_011509701.1 27 - 28 3275 89
XM_011511400.2 XP_011509702.1 27 - 28 3259 89
XM_017004426.1 XP_016859915.1 29 - 30 3536 89
XM_017004427.1 XP_016859916.1 22 - 23 2476 89
XM_017004428.1 XP_016859917.1 28 - 29 3516 89
GenBank mRNA AJ583821.2 - 28 - 29 3444 89
AL833371.1 - 3 - 4 556 89

View Full Product Details