The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018218.2 | NP_060688.1 | 5 - 6 | 733 | 68 |
XM_006712612.3 | XP_006712675.1 | 5 - 6 | 748 | 68 | |
XM_011511396.2 | XP_011509698.1 | 5 - 6 | 748 | 68 | |
XM_011511397.2 | XP_011509699.1 | 6 - 7 | 822 | 68 | |
XM_011511398.2 | XP_011509700.1 | 5 - 6 | 748 | 68 | |
XM_011511400.2 | XP_011509702.1 | 4 - 5 | 533 | 68 | |
XM_011511401.2 | XP_011509703.1 | 5 - 6 | 748 | 68 | |
XM_017004426.1 | XP_016859915.1 | 6 - 7 | 822 | 68 | |
XM_017004428.1 | XP_016859917.1 | 5 - 6 | 632 | 68 | |
GenBank mRNA | AJ583821.2 | - | 5 - 6 | 733 | 68 |
AK001647.1 | - | 5 - 6 | 695 | 68 | |
AK295112.1 | - | 6 - 7 | 815 | 68 | |
BC067300.1 | - | 6 - 7 | 795 | 68 |