The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321449.1 | NP_001308378.1 | 1 - 2 | 315 | 55 |
NM_152358.2 | NP_689571.2 | 1 - 2 | 315 | 55 | |
XM_011526436.2 | XP_011524738.1 | 1 - 2 | 255 | 55 | |
XM_011526437.2 | XP_011524739.1 | 1 - 2 | 270 | 55 | |
GenBank mRNA | AK131024.1 | - | 1 - 2 | 315 | 55 |
AK307321.1 | - | 1 - 2 | 161 | 55 | |
AY358196.1 | - | 1 - 2 | 229 | 55 | |
BC029535.1 | - | 1 - 2 | 196 | 55 |