The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032437.3 | NP_115813.2 | 2 - 3 | 525 | 92 |
XM_005271274.3 | XP_005271331.1 | 2 - 3 | 325 | 92 | |
XM_006710976.3 | XP_006711039.1 | 2 - 3 | 419 | 92 | |
XM_006710977.1 | XP_006711040.1 | 2 - 3 | 447 | 92 | |
XM_011542301.2 | XP_011540603.1 | 2 - 3 | 440 | 92 | |
XM_017002548.1 | XP_016858037.1 | 2 - 3 | 280 | 92 | |
GenBank mRNA | AB058702.1 | - | 2 - 3 | 288 | 92 |
AK092219.1 | - | 2 - 3 | 436 | 92 | |
AK315850.1 | - | 2 - 3 | 301 | 92 | |
BC015814.1 | - | 2 - 3 | 229 | 92 |