The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015302.1 | NP_056117.1 | 12 - 13 | 1068 | 50 |
XM_011526684.2 | XP_011524986.1 | 11 - 12 | 955 | 50 | |
GenBank mRNA | AB020648.1 | - | 12 - 13 | 1041 | 50 |
BC013947.2 | - | 12 - 13 | 1040 | 50 | |
BC064390.1 | - | 11 - 12 | 1004 | 50 | |
BC157882.1 | - | 12 - 13 | 1059 | 50 | |
BC171905.1 | - | 12 - 13 | 1060 | 50 |