The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015302.1 | NP_056117.1 | 16 - 17 | 1573 | 91 |
XM_011526684.2 | XP_011524986.1 | 15 - 16 | 1460 | 91 | |
GenBank mRNA | AB020648.1 | - | 16 - 17 | 1546 | 91 |
BC013947.2 | - | 16 - 17 | 1545 | 91 | |
BC064390.1 | - | 14 - 15 | 1762 | 91 | |
BC157882.1 | - | 16 - 17 | 1564 | 91 | |
BC171905.1 | - | 15 - 16 | 1818 | 91 |