Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr.19: 35612744 - 35625349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015302.1 NP_056117.1 1 - 2 148 68
GenBank mRNA AB020648.1 - 1 - 2 121 68
AK307703.1 - 1 - 2 138 68
BC013947.2 - 1 - 2 120 68
BC064390.1 - 1 - 2 148 68
BC157882.1 - 1 - 2 139 68
BC171905.1 - 1 - 2 140 68

View Full Product Details