The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000613.2 | NP_000604.1 | 3 - 4 | 279 | 125 |
GenBank mRNA | AK303758.1 | - | 3 - 4 | 242 | 125 |
AK313648.1 | - | 3 - 4 | 242 | 125 | |
BC005395.1 | - | 3 - 4 | 279 | 125 | |
J03048.1 | - | 3 - 4 | 215 | 125 | |
X02537.1 | - | 3 - 4 | 155 | 125 |