Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Chromosome Location
Chr.1: 11106531 - 11262551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004958.3 NP_004949.1 13 - 14 2324 66
XM_005263438.2 XP_005263495.1 13 - 14 2331 66
XM_011541166.2 XP_011539468.1 13 - 14 2324 66
XM_017000900.1 XP_016856389.1 10 - 11 1787 66
XM_017000901.1 XP_016856390.1 11 - 12 2075 66
XM_017000902.1 XP_016856391.1 13 - 14 2324 66
GenBank mRNA AB209995.1 - 13 - 14 2305 66
AK302863.1 - 13 - 14 2316 66
AK304273.1 - 13 - 14 2313 66
BC117166.1 - 12 - 13 2221 66
L34075.1 - 13 - 14 2282 66
U88966.1 - 13 - 14 2282 66

View Full Product Details