The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 37 - 38 | 5371 | 67 |
XM_005263438.2 | XP_005263495.1 | 37 - 38 | 5378 | 67 | |
XM_017000900.1 | XP_016856389.1 | 34 - 35 | 4834 | 67 | |
XM_017000901.1 | XP_016856390.1 | 35 - 36 | 5122 | 67 | |
GenBank mRNA | AB209995.1 | - | 37 - 38 | 5352 | 67 |
BC117166.1 | - | 36 - 37 | 5268 | 67 | |
L34075.1 | - | 37 - 38 | 5329 | 67 | |
U88966.1 | - | 37 - 38 | 5326 | 67 |