The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173595.3 | NP_775866.2 | 23 - 24 | 2665 | 55 |
XM_011538197.2 | XP_011536499.1 | 22 - 23 | 2767 | 55 | |
XM_017019183.1 | XP_016874672.1 | 22 - 23 | 2848 | 55 | |
XM_017019184.1 | XP_016874673.1 | 23 - 24 | 2697 | 55 | |
GenBank mRNA | AB190990.1 | - | 23 - 24 | 2682 | 55 |
AK125379.1 | - | 23 - 24 | 2682 | 55 | |
AK126893.1 | - | 16 - 17 | 1825 | 55 | |
BC042070.1 | - | 3 - 4 | 291 | 55 |