The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130146.1 | NP_001123618.1 | 5 - 6 | 632 | 77 |
NM_001130147.1 | NP_001123619.1 | 5 - 6 | 659 | 77 | |
NM_001130148.1 | NP_001123620.1 | 4 - 5 | 605 | 77 | |
NM_032358.3 | NP_115734.1 | 6 - 7 | 686 | 77 | |
GenBank mRNA | AK027638.1 | - | 6 - 7 | 604 | 77 |
AK293165.1 | - | 5 - 6 | 570 | 77 | |
AK316029.1 | - | 4 - 5 | 607 | 77 | |
BC006444.2 | - | 5 - 6 | 575 | 77 | |
CX784327.1 | - | 5 - 6 | 539 | 77 |