Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNJ11
Entrez Gene ID
3767
Gene Name
potassium voltage-gated channel subfamily J member 11
Gene Aliases
BIR, HHF2, IKATP, KIR6.2, MODY13, PHHI, TNDM3
Chromosome Location
Chr.11: 17385246 - 17389331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000525.3 NP_000516.3 1 - 1 1818 144
NM_001166290.1 NP_001159762.1 2 - 2 1163 144
XM_006718226.3 XP_006718289.1 2 - 2 1447 144
XM_017017680.1 XP_016873169.1 2 - 2 1043 144
GenBank mRNA AK301550.1 - 2 - 2 1163 144
BC040617.1 - 2 - 2 1149 144
BC064497.1 - 1 - 1 1817 144
BC112358.1 - 1 - 1 1350 144

View Full Product Details