The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005692.4 | NP_005683.2 | 12 - 13 | 1650 | 76 |
NM_007189.2 | NP_009120.1 | 12 - 13 | 1650 | 76 | |
XM_017011639.1 | XP_016867128.1 | 12 - 13 | 1527 | 76 | |
XM_017011640.1 | XP_016867129.1 | 12 - 13 | 1612 | 76 | |
GenBank mRNA | AF261091.1 | - | 12 - 13 | 1507 | 76 |
AF261092.1 | - | 12 - 13 | 1488 | 76 | |
AJ005016.1 | - | 10 - 11 | 1177 | 76 | |
AL050291.1 | - | 11 - 12 | 1442 | 76 | |
BC001661.2 | - | 12 - 13 | 1482 | 76 | |
BT007451.1 | - | 11 - 12 | 1396 | 76 |