The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145664.1 | NP_001139136.1 | 7 - 8 | 684 | 124 |
XM_011511771.2 | XP_011510073.1 | 8 - 9 | 1222 | 124 | |
XM_011511772.1 | XP_011510074.1 | 10 - 11 | 896 | 124 | |
XM_011511773.1 | XP_011510075.1 | 6 - 7 | 567 | 124 | |
XM_011511776.1 | XP_011510078.1 | 9 - 10 | 855 | 124 | |
XM_011511777.1 | XP_011510079.1 | 4 - 5 | 1225 | 124 | |
XM_011511778.1 | XP_011510080.1 | 4 - 5 | 562 | 124 | |
XM_017004851.1 | XP_016860340.1 | 10 - 11 | 906 | 124 | |
XM_017004852.1 | XP_016860341.1 | 7 - 8 | 693 | 124 | |
XM_017004853.1 | XP_016860342.1 | 10 - 11 | 906 | 124 | |
XM_017004854.1 | XP_016860343.1 | 10 - 11 | 906 | 124 | |
GenBank mRNA | AK298610.1 | - | 7 - 8 | 684 | 124 |