The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017004851.1 | XP_016860340.1 | 1 - 2 | 76 | 77 |
XM_017004852.1 | XP_016860341.1 | 1 - 2 | 76 | 77 | |
XM_017004853.1 | XP_016860342.1 | 1 - 2 | 76 | 77 | |
XM_017004854.1 | XP_016860343.1 | 1 - 2 | 76 | 77 |