The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143821.2 | NP_001137293.2 | 25 - 26 | 3387 | 123 |
NM_001256470.1 | NP_001243399.1 | 29 - 30 | 3711 | 123 | |
NM_001256787.1 | NP_001243716.1 | 23 - 24 | 3267 | 123 | |
NM_019012.5 | NP_061885.2 | 23 - 24 | 3213 | 123 | |
XM_005253400.1 | XP_005253457.1 | 24 - 25 | 3362 | 123 | |
XM_006719091.1 | XP_006719154.1 | 25 - 26 | 3416 | 123 | |
XM_006719093.1 | XP_006719156.1 | 23 - 24 | 3278 | 123 | |
XM_006719096.1 | XP_006719159.1 | 28 - 29 | 3653 | 123 | |
XM_011520714.1 | XP_011519016.1 | 27 - 28 | 3617 | 123 | |
XM_011520715.1 | XP_011519017.1 | 26 - 27 | 3536 | 123 | |
XM_011520716.1 | XP_011519018.1 | 27 - 28 | 3464 | 123 | |
XM_011520717.1 | XP_011519019.1 | 25 - 26 | 3347 | 123 | |
XM_017019495.1 | XP_016874984.1 | 28 - 29 | 3638 | 123 | |
XM_017019496.1 | XP_016874985.1 | 27 - 28 | 3449 | 123 | |
XM_017019497.1 | XP_016874986.1 | 26 - 27 | 3467 | 123 | |
XM_017019498.1 | XP_016874987.1 | 22 - 23 | 3045 | 123 | |
GenBank mRNA | AB051473.2 | - | 25 - 26 | 3283 | 123 |
AB642244.1 | - | 29 - 30 | 3677 | 123 | |
AF302150.1 | - | 23 - 24 | 3111 | 123 | |
AK001529.1 | - | 7 - 8 | 951 | 123 | |
AK023127.1 | - | 11 - 12 | 1234 | 123 | |
AK294739.1 | - | 24 - 25 | 3268 | 123 | |
AL834259.1 | - | 10 - 11 | 1260 | 123 | |
BC004164.2 | - | 14 - 15 | 1898 | 123 | |
BC013133.2 | - | 12 - 13 | 1345 | 123 | |
BC127091.1 | - | 22 - 23 | 3038 | 123 | |
BC127092.1 | - | 23 - 24 | 3111 | 123 |