Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLEKHA5
- Entrez Gene ID
-
54477
- Gene Name
- pleckstrin homology domain containing A5
- Gene Aliases
-
PEPP-2, PEPP2
- Chromosome Location
-
Chr.12: 19129680 - 19376400 on Build GRCh38
View Full Product Details