Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1S
Entrez Gene ID
716
Gene Name
complement component 1, s subcomponent
Gene Aliases
-
Chromosome Location
Chr.12: 7060676 - 7071032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001734.3 NP_001725.1 7 - 8 1153 64
NM_201442.2 NP_958850.1 7 - 8 1240 64
XM_005253760.1 XP_005253817.1 7 - 8 1132 64
GenBank mRNA AK025309.1 - 4 - 5 629 64
AK290295.1 - 7 - 8 1187 64
AM886411.1 - 6 - 7 875 64
BC056903.1 - 7 - 8 1180 64
J04080.1 - 7 - 8 1079 64
M18767.1 - 7 - 8 1099 64

View Full Product Details