The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021822.3 | NP_068594.1 | 4 - 5 | 114 | |
XM_006724290.1 | XP_006724353.1 | 3 - 4 | 114 | ||
XM_017028903.1 | XP_016884392.1 | 4 - 5 | 114 | ||
XM_017028904.1 | XP_016884393.1 | 3 - 4 | 114 | ||
GenBank mRNA | AB266487.1 | - | 4 - 5 | 114 | |
AF182420.1 | - | 4 - 5 | 114 | ||
AK092614.1 | - | 3 - 4 | 1818 | 114 | |
AK310279.1 | - | 4 - 5 | 114 | ||
AK315650.1 | - | 4 - 5 | 114 | ||
BC024268.1 | - | 4 - 5 | 114 | ||
CR456472.1 | - | 4 - 5 | 114 | ||
JF262036.1 | - | 4 - 5 | 114 | ||
JX915801.1 | - | 4 - 5 | 114 |