Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R9A
Entrez Gene ID
55607
Gene Name
protein phosphatase 1 regulatory subunit 9A
Gene Aliases
NRB1, NRBI, Neurabin-I
Chromosome Location
Chr.7: 94907202 - 95296415 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166160.1 NP_001159632.1 16 - 17 3579 117
NM_001166161.1 NP_001159633.1 14 - 15 3393 117
NM_001166162.1 NP_001159634.1 14 - 15 3447 117
XM_011516380.1 XP_011514682.1 17 - 18 3766 117
XM_011516381.1 XP_011514683.1 17 - 18 3642 117
XM_011516382.2 XP_011514684.1 17 - 18 3655 117
XM_011516383.1 XP_011514685.1 17 - 18 3696 117
XM_011516389.1 XP_011514691.1 15 - 16 3513 117
XM_017012394.1 XP_016867883.1 17 - 18 3588 117
XM_017012395.1 XP_016867884.1 17 - 18 3642 117
XM_017012396.1 XP_016867885.1 15 - 16 3637 117
XM_017012397.1 XP_016867886.1 17 - 18 3642 117
XM_017012398.1 XP_016867887.1 15 - 16 3525 117
XM_017012399.1 XP_016867888.1 15 - 16 3459 117
XM_017012400.1 XP_016867889.1 15 - 16 3513 117
XM_017012401.1 XP_016867890.1 16 - 17 3525 117
XM_017012402.1 XP_016867891.1 15 - 16 3459 117
XM_017012403.1 XP_016867892.1 15 - 16 3459 117
XM_017012408.1 XP_016867897.1 15 - 16 1857 117
XM_017012410.1 XP_016867899.1 16 - 17 3645 117
XM_017012411.1 XP_016867900.1 10 - 11 6259 117
GenBank mRNA AK000075.1 - 4 - 5 524 117
AK225204.1 - 4 - 5 524 117
BC130449.1 - 14 - 15 3262 117
BC144108.1 - 14 - 15 3210 118
BC150636.1 - 15 - 16 3527 117

View Full Product Details