The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 85 - 86 | 15072 | 116 |
XM_006721206.2 | XP_006721269.1 | 85 - 86 | 14966 | 116 | |
XM_011523146.2 | XP_011521448.1 | 86 - 87 | 15101 | 116 | |
XM_011523147.1 | XP_011521449.1 | 86 - 87 | 15068 | 116 | |
XM_011523148.1 | XP_011521450.1 | 86 - 87 | 15141 | 116 | |
XM_011523151.1 | XP_011521453.1 | 85 - 86 | 14998 | 116 | |
XM_011523152.1 | XP_011521454.1 | 48 - 49 | 8825 | 116 | |
XM_011523155.2 | XP_011521457.1 | 41 - 42 | 7826 | 116 | |
XM_017023346.1 | XP_016878835.1 | 85 - 86 | 15244 | 116 | |
XM_017023347.1 | XP_016878836.1 | 73 - 74 | 13483 | 116 | |
XM_017023348.1 | XP_016878837.1 | 73 - 74 | 13223 | 116 | |
GenBank mRNA | AK026688.1 | - | 6 - 7 | 1168 | 116 |
AK074472.1 | - | 10 - 11 | 1891 | 116 | |
AL137259.1 | - | 13 - 14 | 2468 | 116 | |
BC043273.2 | - | 21 - 22 | 3916 | 116 |