Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSBP2
Entrez Gene ID
23635
Gene Name
single stranded DNA binding protein 2
Gene Aliases
HSPC116, SOSS-B2
Chromosome Location
Chr.5: 81417360 - 81751253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256732.1 NP_001243661.1 15 - 16 1187 75
NM_001256733.1 NP_001243662.1 14 - 15 1103 75
NM_001256734.1 NP_001243663.1 14 - 15 1097 75
NM_001256735.1 NP_001243664.1 14 - 15 1073 75
NM_012446.3 NP_036578.2 15 - 16 1163 75
XM_017009304.1 XP_016864793.1 15 - 16 1005 75
XM_017009305.1 XP_016864794.1 16 - 17 1094 75
XM_017009307.1 XP_016864796.1 14 - 15 891 75
XM_017009308.1 XP_016864797.1 16 - 17 1266 75
XM_017009309.1 XP_016864798.1 15 - 16 1330 75
XM_017009310.1 XP_016864799.1 15 - 16 1478 75
GenBank mRNA AF077048.1 - 15 - 16 1027 75
AF161465.1 - 15 - 16 993 75
AK074537.1 - 15 - 16 958 75
AK293544.1 - 14 - 15 898 75
AK297494.1 - 15 - 16 871 75
AK299858.1 - 14 - 15 941 75
AK312337.1 - 15 - 16 1007 75
AL080076.1 - 15 - 16 1011 75
BC017020.1 - 15 - 16 1011 75
BX397789.2 - 2 - 3 75
CR936787.1 - 14 - 15 918 75
DA202987.1 - 7 - 8 353 75

View Full Product Details