The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003737.3 | NP_003728.1 | 3 - 4 | 2392 | 129 |
GenBank mRNA | AB053446.1 | - | 3 - 4 | 2395 | 129 |