The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040274.2 | NP_001035364.2 | 14 - 15 | 1366 | 90 |
GenBank mRNA | AM293674.1 | - | 14 - 15 | 1085 | 90 |
BC012225.1 | - | 4 - 5 | 263 | 90 | |
DA620756.1 | - | 5 - 6 | 340 | 90 |