Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 24 - 25 5406 98
XM_011523663.1 XP_011521965.1 26 - 27 4274 98
XM_011523664.2 XP_011521966.1 27 - 28 4423 98
XM_011523666.1 XP_011521968.1 26 - 27 4274 98
XM_011523667.2 XP_011521969.1 25 - 26 4177 98
XM_011523668.1 XP_011521970.1 16 - 17 2657 98
XM_011523669.2 XP_011521971.1 14 - 15 2341 98
XM_011523670.2 XP_011521972.1 26 - 27 4274 98
XM_017024218.1 XP_016879707.1 13 - 14 2358 98
XM_017024219.1 XP_016879708.1 26 - 27 4274 98
GenBank mRNA AB040936.2 - 24 - 25 5406 98
BX648163.1 - 6 - 7 996 98

View Full Product Details