The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252385.1 | NP_001239314.1 | 13 - 14 | 1639 | 64 |
NM_001252386.1 | NP_001239315.1 | 12 - 13 | 1467 | 64 | |
NM_001252390.1 | NP_001239319.1 | 13 - 14 | 1504 | 64 | |
NM_001252391.1 | NP_001239320.1 | 13 - 14 | 1536 | 64 | |
NM_001252392.1 | NP_001239321.1 | 13 - 14 | 1536 | 64 | |
NM_001252393.1 | NP_001239322.1 | 13 - 14 | 1639 | 64 | |
NM_001258454.1 | NP_001245383.1 | 13 - 14 | 1639 | 64 | |
NM_001258455.1 | NP_001245384.1 | 12 - 13 | 1434 | 64 | |
NM_001258456.1 | NP_001245385.1 | 12 - 13 | 1434 | 64 | |
NM_006058.4 | NP_006049.3 | 13 - 14 | 1639 | 64 | |
XM_005268355.1 | XP_005268412.1 | 13 - 14 | 1552 | 64 | |
XM_006714751.1 | XP_006714814.1 | 13 - 14 | 1552 | 64 | |
XM_006714752.2 | XP_006714815.1 | 13 - 14 | 1552 | 64 | |
XM_017008945.1 | XP_016864434.1 | 13 - 14 | 1552 | 64 | |
XM_017008946.1 | XP_016864435.1 | 13 - 14 | 1518 | 64 | |
XM_017008947.1 | XP_016864436.1 | 14 - 15 | 2021 | 64 | |
XM_017008948.1 | XP_016864437.1 | 12 - 13 | 1813 | 64 | |
XM_017008949.1 | XP_016864438.1 | 13 - 14 | 1552 | 64 | |
XM_017008950.1 | XP_016864439.1 | 13 - 14 | 1552 | 64 | |
XM_017008951.1 | XP_016864440.1 | 12 - 13 | 1813 | 64 | |
GenBank mRNA | AB177543.1 | - | 12 - 13 | 1398 | 64 |
AB177544.1 | - | 12 - 13 | 1398 | 64 | |
AB252970.1 | - | 13 - 14 | 1508 | 64 | |
AB252971.1 | - | 13 - 14 | 1508 | 64 | |
AB252972.1 | - | 13 - 14 | 1629 | 64 | |
AB252973.1 | - | 13 - 14 | 1629 | 64 | |
AB252976.1 | - | 11 - 12 | 1262 | 64 | |
AB252977.1 | - | 11 - 12 | 1262 | 64 | |
AB252978.1 | - | 11 - 12 | 1262 | 64 | |
AB252979.1 | - | 11 - 12 | 1262 | 64 | |
AB252980.1 | - | 11 - 12 | 1262 | 64 | |
AB252981.1 | - | 11 - 12 | 1262 | 64 | |
AB252982.1 | - | 13 - 14 | 1508 | 64 | |
AJ011895.1 | - | 13 - 14 | 1508 | 64 | |
AJ011896.1 | - | 13 - 14 | 1508 | 64 | |
AK074400.1 | - | 13 - 14 | 1570 | 64 | |
AK290999.1 | - | 13 - 14 | 1505 | 64 | |
AK298685.1 | - | 12 - 13 | 1461 | 64 | |
AK299975.1 | - | 12 - 13 | 1399 | 64 | |
AK303573.1 | - | 13 - 14 | 1589 | 64 | |
AY012155.1 | - | 13 - 14 | 1655 | 64 | |
BC012133.1 | - | 13 - 14 | 1545 | 64 | |
BC014008.1 | - | 13 - 14 | 1486 | 64 | |
BX640647.1 | - | 12 - 13 | 1388 | 64 | |
D30755.2 | - | 12 - 13 | 1398 | 64 | |
U39403.1 | - | 9 - 10 | 994 | 64 |