The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252386.1 | NP_001239315.1 | 15 - 16 | 1852 | 82 |
NM_001252390.1 | NP_001239319.1 | 16 - 17 | 1889 | 82 | |
NM_001252391.1 | NP_001239320.1 | 16 - 17 | 1921 | 82 | |
NM_001252392.1 | NP_001239321.1 | 16 - 17 | 1921 | 82 | |
NM_001252393.1 | NP_001239322.1 | 16 - 17 | 2024 | 82 | |
NM_001258454.1 | NP_001245383.1 | 16 - 17 | 2024 | 82 | |
NM_006058.4 | NP_006049.3 | 16 - 17 | 2024 | 82 | |
XM_017008945.1 | XP_016864434.1 | 16 - 17 | 1937 | 82 | |
XM_017008946.1 | XP_016864435.1 | 16 - 17 | 1903 | 82 | |
XM_017008947.1 | XP_016864436.1 | 17 - 18 | 2406 | 82 | |
XM_017008948.1 | XP_016864437.1 | 15 - 16 | 2198 | 82 | |
XM_017008951.1 | XP_016864440.1 | 15 - 16 | 2198 | 82 | |
GenBank mRNA | AB252970.1 | - | 16 - 17 | 1893 | 82 |
AB252971.1 | - | 16 - 17 | 1893 | 82 | |
AB252972.1 | - | 16 - 17 | 2014 | 82 | |
AB252973.1 | - | 16 - 17 | 2014 | 82 | |
AB252982.1 | - | 16 - 17 | 1893 | 82 | |
AJ011895.1 | - | 16 - 17 | 1893 | 82 | |
AJ011896.1 | - | 16 - 17 | 1893 | 82 | |
AK074400.1 | - | 16 - 17 | 1955 | 82 | |
AK290999.1 | - | 16 - 17 | 1890 | 82 | |
AK298685.1 | - | 15 - 16 | 1846 | 82 | |
AK299975.1 | - | 15 - 16 | 1784 | 82 | |
AY012155.1 | - | 16 - 17 | 2040 | 82 | |
BC012133.1 | - | 16 - 17 | 1930 | 82 | |
BC014008.1 | - | 16 - 17 | 1871 | 82 | |
BX640647.1 | - | 15 - 16 | 1773 | 82 | |
D30755.2 | - | 15 - 16 | 1783 | 82 | |
U39403.1 | - | 12 - 13 | 1379 | 82 |