The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252385.1 | NP_001239314.1 | 3 - 4 | 510 | 70 |
NM_001252386.1 | NP_001239315.1 | 2 - 3 | 338 | 70 | |
NM_001252390.1 | NP_001239319.1 | 3 - 4 | 375 | 70 | |
NM_001252391.1 | NP_001239320.1 | 3 - 4 | 407 | 70 | |
NM_001252392.1 | NP_001239321.1 | 3 - 4 | 407 | 70 | |
NM_001252393.1 | NP_001239322.1 | 3 - 4 | 510 | 70 | |
NM_001258454.1 | NP_001245383.1 | 3 - 4 | 510 | 70 | |
NM_001258455.1 | NP_001245384.1 | 2 - 3 | 305 | 70 | |
NM_001258456.1 | NP_001245385.1 | 2 - 3 | 305 | 70 | |
NM_006058.4 | NP_006049.3 | 3 - 4 | 510 | 70 | |
XM_005268355.1 | XP_005268412.1 | 3 - 4 | 423 | 70 | |
XM_006714751.1 | XP_006714814.1 | 3 - 4 | 423 | 70 | |
XM_006714752.2 | XP_006714815.1 | 3 - 4 | 423 | 70 | |
XM_017008945.1 | XP_016864434.1 | 3 - 4 | 423 | 70 | |
XM_017008946.1 | XP_016864435.1 | 3 - 4 | 389 | 70 | |
XM_017008947.1 | XP_016864436.1 | 4 - 5 | 892 | 70 | |
XM_017008948.1 | XP_016864437.1 | 2 - 3 | 684 | 70 | |
XM_017008949.1 | XP_016864438.1 | 3 - 4 | 423 | 70 | |
XM_017008950.1 | XP_016864439.1 | 3 - 4 | 423 | 70 | |
XM_017008951.1 | XP_016864440.1 | 2 - 3 | 684 | 70 | |
GenBank mRNA | AB177543.1 | - | 2 - 3 | 269 | 70 |
AB177544.1 | - | 2 - 3 | 269 | 70 | |
AB252970.1 | - | 3 - 4 | 379 | 70 | |
AB252971.1 | - | 3 - 4 | 379 | 70 | |
AB252972.1 | - | 3 - 4 | 500 | 70 | |
AB252973.1 | - | 3 - 4 | 500 | 70 | |
AB252976.1 | - | 1 - 2 | 133 | 70 | |
AB252977.1 | - | 1 - 2 | 133 | 70 | |
AB252978.1 | - | 1 - 2 | 133 | 70 | |
AB252979.1 | - | 1 - 2 | 133 | 70 | |
AB252980.1 | - | 1 - 2 | 133 | 70 | |
AB252981.1 | - | 1 - 2 | 133 | 70 | |
AB252982.1 | - | 3 - 4 | 379 | 70 | |
AJ011895.1 | - | 3 - 4 | 379 | 70 | |
AJ011896.1 | - | 3 - 4 | 379 | 70 | |
AK074400.1 | - | 3 - 4 | 441 | 70 | |
AK290999.1 | - | 3 - 4 | 376 | 70 | |
AK298685.1 | - | 2 - 3 | 332 | 70 | |
AK299975.1 | - | 2 - 3 | 270 | 70 | |
AK303573.1 | - | 3 - 4 | 460 | 70 | |
AY012155.1 | - | 3 - 4 | 526 | 70 | |
BC012133.1 | - | 3 - 4 | 416 | 70 | |
BC014008.1 | - | 3 - 4 | 357 | 70 | |
BP254790.1 | - | 3 - 4 | 500 | 70 | |
BX640647.1 | - | 2 - 3 | 259 | 70 | |
D30755.2 | - | 2 - 3 | 269 | 70 |