The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323281.1 | NP_001310210.1 | 8 - 9 | 1613 | 139 |
NM_001323282.1 | NP_001310211.1 | 9 - 10 | 1678 | 139 | |
NM_002895.4 | NP_002886.2 | 9 - 10 | 1540 | 139 | |
NM_183404.3 | NP_899662.1 | 9 - 10 | 1540 | 139 | |
XM_006723842.2 | XP_006723905.1 | 8 - 9 | 1251 | 139 | |
XM_011528955.2 | XP_011527257.1 | 9 - 10 | 1316 | 139 | |
XM_011528958.2 | XP_011527260.1 | 9 - 10 | 1316 | 139 | |
XM_017027992.1 | XP_016883481.1 | 10 - 11 | 1496 | 139 | |
GenBank mRNA | AK290380.1 | - | 9 - 10 | 1316 | 139 |
AK308096.1 | - | 9 - 10 | 1301 | 139 | |
BC032247.1 | - | 9 - 10 | 1323 | 139 | |
L14812.1 | - | 9 - 10 | 1321 | 139 | |
M74547.1 | - | 7 - 8 | 854 | 139 |