The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319044.1 | NP_001305973.1 | 2 - 3 | 220 | 95 |
NM_001319045.1 | NP_001305974.1 | 2 - 3 | 220 | 95 | |
NM_001319046.1 | NP_001305975.1 | 2 - 3 | 220 | 95 | |
NM_032025.4 | NP_114414.2 | 2 - 3 | 220 | 95 | |
XM_011513224.1 | XP_011511526.1 | 2 - 3 | 203 | 95 | |
GenBank mRNA | AF109358.1 | - | 1 - 2 | 77 | 95 |
AF172818.1 | - | 1 - 2 | 88 | 95 | |
AF212241.1 | - | 2 - 3 | 169 | 95 | |
AF497978.1 | - | 2 - 3 | 98 | 95 | |
AK027356.1 | - | 2 - 3 | 111 | 95 | |
AK293993.1 | - | 2 - 3 | 111 | 95 | |
AL833101.1 | - | 2 - 3 | 104 | 95 | |
BC011885.1 | - | 2 - 3 | 101 | 95 | |
BX647951.1 | - | 2 - 3 | 105 | 95 | |
DA382303.1 | - | 2 - 3 | 221 | 95 |