The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002180.2 | NP_002171.2 | 3 - 4 | 557 | 104 |
XM_005273974.2 | XP_005274031.1 | 3 - 4 | 521 | 104 | |
XM_005273976.1 | XP_005274033.1 | 3 - 4 | 557 | 104 | |
XM_017017671.1 | XP_016873160.1 | 3 - 4 | 557 | 104 | |
GenBank mRNA | AB208802.1 | - | - | 499 | 104 |
AK312565.1 | - | 3 - 4 | 515 | 104 | |
BC000290.2 | - | 3 - 4 | 470 | 104 | |
BC025299.1 | - | 3 - 4 | 502 | 104 | |
BC080519.1 | - | 3 - 4 | 502 | 104 | |
BC105088.1 | - | 3 - 4 | 494 | 104 | |
BC105090.1 | - | 3 - 4 | 494 | 104 | |
L14754.1 | - | 3 - 4 | 504 | 104 |